EMAIL THIS PAGE TO A FRIEND
Meditsina truda i promyshlennaia ekologiia
[Methyl sulphides and their metabolites in biologic materials of workers engaged into sulfate pulping].
PMID 17695266
Abstract
The authors studied serum and urine levels of methyl sulphides in workers engaged into sulfate pulping and exposed to methyl sulphides. Blood serum was studied only for dimethyldisulfide. Urine appeared to contain no methyl sulphides, but finding was increased urinary excretion of sulfates which are metabolites of methyl sulphides. Sulfates content of urine correlated with intensity and duration of exposure to methyl sulphides, so this exposure test could be used as a biologic marker in monitoring work conditions and health state of workers exposed to methyl sulphides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
318183
Methyl sulfate sodium salt
CH3NaO4S