EMAIL THIS PAGE TO A FRIEND
American journal of hematology
Malaria and hereditary elliptocytosis.
PMID 17696197
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q5001
Quinidine gluconate salt, analytical standard
C20H24N2O2 · C6H12O7