EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Microbial transformation of neoandrographolide by Aspergillus niger (AS 3.739).
PMID 17701567
Abstract
The biotransformation of neoandrographolide (1) was investigated by using Aspergillus niger (AS 3.739). Five products were obtained and identified as 8(17),13-ent-labdadien-16,15-olid-19-oic acid (2), 19-hydroxy-8(17),13-ent-labdadien-16,15-olide (3), 18-hydroxy-8(17),13-ent-labdadien-16,15-olid-19-oic acid (4), 3alpha-hydroxy-8(17),13-ent-labdadien-16,15-olid-19-oic acid (5) and 8beta,19-dihydroxy-ent-labd-13-en-16,15-olide (6) by spectroscopic and chemical means. Products 4, 5 and 6 are new compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
49879
Neoandrographolide, analytical standard
C26H40O8