EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
On the synthesis of protopine alkaloids.
PMID 17705430
Abstract
For the synthesis of protopine alkaloids, we studied a reaction sequence based on a ring enlargement of indeno[2,1-a][3]benzazepines by a singlet oxygen oxygenation, followed by conversion of an amide carbonyl group of the resultant 10-membered keto-lactam to a methylene group, which is the last step for completion of the synthesis. The key substances, indeno[2,1-a][3]benzazepines, were prepared by Bischler-Napieralski cyclization of alkoxy-substituted 1-(2-bromobenzyl)-3-benzazepin-2-ones. Steric effects of the substituents in this synthesis were examined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8489
Protopine hydrochloride, ≥98%, solid
C20H19NO5·HCl