EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Cloud point formation based on mixed micelle in the presence of electrolyte for extraction, preconcentration, and spectrophotometric determination of trace amounts of hydrazine in water and biological samples.
PMID 17706585
Abstract
A cloud point extraction process using mixed micelle of the anionic surfactant sodium dodecyl sulfate and the nonionic surfactant Triton X-114 to extract hydrazine from aqueous solutions was investigated. The method is based on the condensation reaction of hydrazine with p-(dimethylamino)benzaldehyde, azine formation, and mixed micelle-mediated extraction of azine in the presence of NaCl electrolyte as an inducing phase separation. An azine product was concentrated in surfactant-rich phase after separation. The optimal extraction and reaction conditions (e.g., surfactant, reagent and electrolyte concentrations, and centrifuge time) were studied and the analytical characteristics of the method (e.g., limit of detection, linear range, preconcentration, and improvement factors) were obtained. Linearity was obeyed in the range of 0.50-110ngml(-1) of hydrazine and the detection limit of the method is 0.08ngml(-1). The interference effect of some cations, anions, and organic compounds was also tested. The method was successfully applied to the determination of hydrazine in water and biological samples.