EMAIL THIS PAGE TO A FRIEND
La Medicina del lavoro
[A case of methemoglobinemia due to 4-nitrobenzonitrile exposure].
PMID 1770871
Abstract
The paper describes a singular case of acute intoxication by 4-nitrobenzonitrile, a rare intermediate compound produced by pharmaceutical synthesis. The patients subsequently showed an increase in triglycerides and fatty alterations of the liver.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N12007
4-Nitrobenzonitrile, 97%
C7H4N2O2