EMAIL THIS PAGE TO A FRIEND
Neuropediatrics
Lessons to learn from rare inborn errors of metabolism.
PMID 17712731
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
102687
Ethylmalonic acid, 97%
C5H8O4