EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Cannabidiol--recent advances.
PMID 17712814
Abstract
The aim of this review is to present some of the recent publications on cannabidiol (CBD; 2), a major non-psychoactive constituent of Cannabis, and to give a general overview. Special emphasis is laid on biochemical and pharmacological advances, and on novel mechanisms recently put forward, to shed light on some of the pharmacological effects that can possibly be rationalized through these mechanisms. The plethora of positive pharmacological effects observed with CBD make this compound a highly attractive therapeutic entity.