EMAIL THIS PAGE TO A FRIEND
VASA. Supplementum
[A new oral prostaglandin E1 derivative (OP 1206) compared with intravenous PGE1: assessment of the effect on macro- and microcirculation].
PMID 1771541
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6538
Limaprost, ≥99%, crystalline
C22H36O5