EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Lewis acid catalyzed propargylation of arenes with O-propargyl trichloroacetimidates: synthesis of 1,3-diarylpropynes.
PMID 17715969
Abstract
The BF3.OEt2-catalyzed Friedel-Crafts propargylation of aromatic compounds with O-propargyl trichloroacetimidates is highly efficient and affords 1,3-diarylpropyne derivatives in good yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480983
Propyne, ≥97%
C3H4
295493
Propyne, ≥99%
C3H4