EMAIL THIS PAGE TO A FRIEND
Environmental entomology
Effects of the insecticide phosmet on solitary bee foraging and nesting in orchards of Capitol Reef National Park, Utah.
PMID 17716471
Abstract
Capitol Reef National Park, in southcentral Utah, contains 22 small orchards planted with antique fruit varieties by Mormon pioneers beginning over a century ago. The orchards continue to be managed in a pick-and-pay program, which includes spraying with phosmet to suppress codling moth (Cydia pomonella L.). The park is also home to a rich diversity of flowering plants, many of which are rare, bee-pollinated, and have populations within 1 km of the orchards. Over 3 yr, we studied the short-term effects of phosmet spraying on bee populations: (1) foraging on plants within the orchard understory and adjacent to it; and (2) nesting in, and at several distances from, the orchards. We recorded a rich bee fauna (47 taxa) in the orchards and on plants nearby. In 2 yr (2002 and 2004), we found no difference in the number of native bee visits to several species of plants flowering in and near to orchards immediately before and 1 d after spraying. Conversely, our nesting studies using the semidomesticated alfalfa leafcutting bee, Megachile rotundata (F.), showed strong significant declines in the number of adult males, nesting females, and progeny production subsequent to spraying at distances up to 160 m from sprayed orchards where the bees were presumably foraging. We showed that M. rotundata is negatively affected by phosmet spraying and suggest that caution should be exercised in its use in areas where bees are apt to forage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36195
Phosmet, PESTANAL®, analytical standard
C11H12NO4PS2