EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Analysis of phenolic acids as chloroformate derivatives using solid phase microextraction-gas chromatography.
PMID 17723529
Abstract
In the presented study, a simple and original procedure of phenolic acids derivatization treated by ethyl and methyl chloroformate performed in an aqueous media consisting of acetonitrile, water, methanol/ethanol and pyridine has been modified and optimized. Seven phenolic acid standards-caffeic, ferulic, gallic, p-coumaric, protocatechuic, syringic and vanillic were derivatized into corresponding methyl/ethyl esters and subsequently determined by the means of gas chromatography connected to the flame-ionisation detector (FID). Some selected validation parameters as linearity, detection and quantitation limits and peak area repeatability were valued. The total time of gas chromatography (GC) analysis was 24 min for methyl chloroformate and 30 min for ethyl chloroformate derivatization. The more suitable methyl chloroformate derivatization was used for further experiments on the possibility of multiple pre-concentration by the direct solid phase microextraction technique (SPME). For this purpose, polyacrylate (PA), polydimethylsiloxane (PDMS), carboxen/polydimethylsiloxane (CAR/PDMS) and polydimethylsiloxane/divinylbenzene (PDMS/DVB) fibres were tested and the extraction conditions concerning time of extraction, temperature and time of desorption were optimized. The most polar PA fibre gave the best results under optimal extraction conditions (50 min extraction time, 25 degrees C extraction temperature and 10 min desorption time). As a result, the total time of SPME-GC analysis was 74 min and an increase in method sensitivity was reached. The limits of quantitation (LOQ) of p-coumaric, ferulic, syringic and vanillic acid esters after SPME pre-concentration were 0.02, 0.17, 0.2 and 0.2 microg mL(-1), respectively, showing approximately 10 times higher sensitivity in comparison with the original GC method.