EMAIL THIS PAGE TO A FRIEND
Journal of peptide science : an official publication of the European Peptide Society
Fast Fmoc synthesis of hAmylin1-37 with pseudoproline assisted on-resin disulfide formation.
PMID 17726722
Abstract
Human amylin (1-37) and the (1-13) fragment were synthesized with and without pseudoproline dipeptides. Thallium (III) trifluoroacetate, a mild oxidant, was used to cyclize the peptides by forming a disulfide bridge from C(2) to C(7). On the basis of our model studies, incorporation of a pseudoproline dipeptide decreases the amount of time necessary for the crude linear amylin (1-13) to cyclize on the resin. Without pseudoproline dipeptides, the 1-37 crude linear amylin was not pure enough to undergo the cyclization reaction. Following the cyclization studies, the synthesis time of the linear human amylin (1-37) was systematically reduced from 58 h to 8.5 h by shortening the reaction times. Cyclization and cleavage times were also reduced to 1.5 h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
150533
Thallium(III) trifluoroacetate, technical grade
C6F9O6Tl