EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Identification and characterization of several dietary alkaloids as weak inhibitors of hedgehog signaling.
PMID 17728282
Abstract
The Hedgehog (Hh) signaling pathway plays an integral role in the patterning and development of diverse structures in the vertebrate embryo. Aberrations in Hh signaling are associated with a range of developmental defects including failure of interhemispheric division of the embryonic forebrain as well as midline facial dysmorphia including cleft lip/palate and cyclopia, collectively termed holoprosencephaly (HPE). Postnatally, Hh signaling has been postulated to play a pivotal role in healing and repair processes and inappropriate Hh pathway activation has been implicated in several types of cancers. The Veratrum alkaloid cyclopamine is a potent inhibitor of Hh signaling and causes HPE-like defects in diverse species including sheep, hamster, mouse, and zebra fish. Using murine cell-based assays, we have determined that a number of dietary alkaloids similar in structure to cyclopamine also inhibit Hh signaling but with significantly lower potency. We found that these dietary compounds act additively through a mechanism similar to cyclopamine, downstream of Ptc1 and upstream of Gli1. Using an embryonic zebra fish developmental assay, we found that while cyclopamine exposure caused HPE-like defects, exposure to one of these dietary compounds, solanidine, did not.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13264
Solanidine, ≥97.0% (HPLC)
C27H43NO