EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The discovery of 6-amino nicotinamides as potent and selective histone deacetylase inhibitors.
PMID 17761416
Abstract
This communication highlights the development of a nicotinamide series of histone deacetylase inhibitors within the benzamide structural class. Extensive exploration around the nicotinamide core led to the discovery of a class I selective HDAC inhibitor that possesses excellent intrinsic and cell-based potency, acceptable ancillary pharmacology, favorable pharmacokinetics, sustained pharmacodynamics in vitro, and achieves in vivo efficacy in an HCT116 xenograft model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A68203
6-Aminonicotinamide, 99%
C6H7N3O