EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Dissipation of fenoxycarb and pyriproxyfen in fresh and canned peach.
PMID 17763032
Abstract
The objective of this work was to determine the dissipation of fenoxycarb and pyriproxyfen in fresh and canned peaches in order to know the levels of residues that can reach consumers in real circumstances. Two field dissipation studies were carried out, one of them at the pre-harvest interval (PHI) with good agricultural practice (GAP) and the other one in a situation of critical agricultural practice (CAP). Two canning dissipation studies were carried out for samples from both agricultural situations in an industrial pilot plant and the dissipation was determined in each relevant step. An analytical methodology was used including acetone-dichloromethane extraction, purification and analysis by liquid chromatography and diode array detection (LC-DAD) with a limit of quantification (LOQ) of 0.05 mg/kg. It was validated under SANCO/10232/2006 Guidelines. These pesticides complied with the official maximum residue limits (MRLs) in peaches at the PHI with good agricultural practices. In hypothetical situation of a second application at the PHI, fenoxycarb and pyriproxyfen residues were above the MRLs in peaches. The canning study reduced the residues to no detectable levels in the cans for consumers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34343
Fenoxycarb, PESTANAL®, analytical standard
C17H19NO4