EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Assessing supercritical fluid extraction for the analysis of fipronil, kresoxim-methyl, acrinathrin, and pyridaben residues in melon.
PMID 17763038
Abstract
An analytical method, using supercritical fluid extraction (SFE), was assessed for residue analysis of the insecticides fipronil, acrinathrin and pyridaben and the fungicide kresoxim-methyl in melons. Extraction conditions were optimized on glass beads, as inert material, evaluating different pressures, temperatures, and CO(2) volumes. Also, the extractability of pesticides from hydromatrix and anhydrous magnesium sulfate was evaluated. Both adsorbents were evaluated for the preparation of SFE-spiked melon samples. Extractions of fortified samples were done using 20 mL of CO(2), 300 atm, and 50 degrees C. Different extraction conditions, including the addition of water and methanol as a static modifier, were used to assess the extractability of these pesticides, and the addition of methanol was demonstrated to improve the extraction of all the studied pesticides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46415
Acrinathrin, PESTANAL®, analytical standard
C26H21F6NO5