EMAIL THIS PAGE TO A FRIEND
Renal failure
Do AshSplit haemodialysis catheters provide better flow rates in the long term?
PMID 17763168
Abstract
Recently, interventional radiologists have adopted an increasingly prominent role in the placement and management of hemodialysis catheters, as well as in the research and development of new and better catheters. The purpose of this study was to evaluate the viability and hemodialysis efficiency of the AshSplit catheter and the Permcath catheter. 204 consecutive patients requiring radiological insertion of hemodialysis catheters were followed, retrospectively, over a 42-month period. Both hemodialysis catheters were placed using a combination of ultrasonic and fluoroscopic guidance and tunneled appropriately. Information collected included catheter insertion sites, insertion complications, catheter duration, and final outcome. Over the study period of two years, 269 catheters were placed into 204 patients with end stage renal failure. Patients received either an AshSplit (101 patients, 127 catheters) or a Permcath (103 patients, 142 catheters). Vascular access route of choice was the right internal jugular vein (67% AshSplit, 71% Permcath). Insertion complications occurred in 18 patients overall (6.6%), with only 1 requiring further intervention (hemopneumothorax). Flow rates averaged 259 mls/min for AshSplits and 248 mls/min for Permcaths (p < 0.001). Follow-up of catheter viability for 42 months yielded a mean AshSplit catheter duration of 246 days (range 6-932) and 239 days (range 1-1,278) for Permcath (p = 0.46). Reasons for catheter failure and elective catheter removal were similar in both groups; however, Permcaths required significantly more thrombolysis than AshSplits, p < 0.001. The AshSplit provides significantly better flow rates and less thrombolysis compared to the Permcath, with similar catheter dwell times.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7263
Ethynodiol diacetate
C24H32O4