EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Rapid simultaneous determination of 14 sulfonamides in wastewater by liquid chromatography tandem mass spectrometry.
PMID 17763515
Abstract
This paper describes a rapid method for the determination of 14 kinds of sulfonamides (SAs) in wastewater using SPE, and LC-MS/MS with positive ESI (ESI(+)) and selected reaction monitoring (SRM) mode. The SPE was performed on an Oasis hydrophilic-lipophilic-balanced (HLB) cartridge. Chromatographic separation on a C18 column was achieved using a binary eluent containing methanol and water with 0.2% formic acid. Typical recoveries of the analytes ranged from 22.3 to 87.0% at a fortification level of 100 ng/L. The LODs in wastewater except sulfathiazole (3 ng/L) could be detected and quantified at levels as low as 1 ng/L. Finally, the method was applied to water from the municipal outlet and the aquaculture wastewater effluent. Sulfamethazine (SM(2)), sulfamethoxypyridazine (SMP), and sulfamethoxazole (SMZ) were most frequently found in wastewater in a concentration range between 1.2 and 31.7 ng/L.
Related Materials