EMAIL THIS PAGE TO A FRIEND
Life sciences
Adenosine and TNF-alpha exert similar inotropic effect on heart cultures, suggesting a cardioprotective mechanism against hypoxia.
PMID 17764703
Abstract
When cardiomyocytes were subjected to hypoxia, tumor necrosis factor-alpha (TNF-alpha; 3-50 ng/ml) or adenosine (1-100 microM), decreased hypoxic damage as was detected by lactate dehydrogenase (LDH) release, MTT (3-[4,5-Dimethylthiazol-2-yl]-2,5-diphenyltetrazolium bromide) absorbance, ROS (reactive oxygen species) measurement or desmin immunostaining. This cardioprotection was not prevented in TNF-alpha-treated cultures by 5-hydroxydecanoic acid (5-HD). Our aim was to elucidate whether adenosine and TNF-alpha mediate a similar protective mechanism against hypoxia in primary heart cultures and in H9c2 cardiomyocytes. Adenosine and TNF-alpha are known for their negative inotropic effects on the heart. We have suggested that deoxyglucose uptake reflects heart contractility in cell cultures; therefore, we assayed its accumulation under various conditions. Treatment for 20 min with adenosine, R-PIA [(-)-N(6)-phenylisopropyladenosine] (10 microM), or TNF-alpha reduced (3)H-deoxyglucose uptake in primary heart cultures and also in H9c2 cardiomyocytes by 30-50%. Isoproterenol accelerated (3)H-deoxyglucose uptake by 50%. Adenosine, R-PIA, or TNF-alpha attenuated the stimulatory effect of isoproterenol on (3)H-deoxyglucose uptake to control levels. Hypoxia reduced (3)H-deoxyglucose uptake by 50%, as in the treatment of the hypoxic cultures with TNF-alpha or adenosine. Glibenclamide (2 microM), 5-HD (300 microM), or diazoxide (50 microM) increased (3)H-deoxyglucose uptake by 50-80%. Adenosine (100 microM) and TNF-alpha (50 ng/ml) stimulated (86)Rb efflux. Glibenclamide attenuated this effect. We demonstrate that TNF-alpha, like adenosine, accelerated Ca(2+) uptake into the sarcoplasmic reticulum (SR) by 50-100% and therefore prevented cardiomyocyte Ca(2+) overload. Our findings further suggest that TNF-alpha, as well as adenosine, may mediate an adaptive effect in the heart by preventing Ca(2+) overload via activation of SR Ca-ATPase (SERCA(2)a).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4532
(−)-N6-(2-Phenylisopropyl)adenosine, solid
C19H23N5O4