EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Receptor mediation of exaggerated responses to serotonin-enhancing drugs in serotonin transporter (SERT)-deficient mice.
PMID 17765930
Abstract
Administration of serotonin-enhancing drugs induces a distinctive behavioral syndrome in rodents. We previously reported that mice with a targeted disruption of the serotonin transporter (SERT) display some of these behaviors spontaneously, in the absence of drug. In the current studies, we assessed the drug-induced serotonin syndrome in SERT wildtype (+/+), heterozygous (+/-) and knockout (-/-) mice. In SERT -/- mice, the monoamine oxidase inhibitor (MAOI) tranylcypromine (1mg/kg) or the serotonin precursor 5-hydroxy-L-tryptophan (5-HTP; 80 mg/kg) led to markedly exaggerated serotonin syndrome behaviors relative to SERT +/+ mice, with an intermediate phenotype in SERT +/- mice. SERT +/+ mice developed significant serotonin syndrome behaviors only with the combination of the MAO-A/B inhibitor tranylcypromine (0.5 or 1 mg/kg) or the MAO-A-selective inhibitor clorgyline (1.2 mg/kg) plus 5-HTP. In evaluations of underlying mechanisms, pretreatment with the Htr1a receptor antagonist WAY 100635 (1 mg/kg), but not the Htr7 antagonist SB 269970 (3 mg/kg) or the Htr2a antagonist MDL 11,939 (5 mg/kg), markedly decreased the exaggerated 5-HTP-induced behaviors in SERT -/- mice. Subsequent experiments showed that the Htr1a agonist 8-OH-DPAT (1 or 2 mg/kg) elicited serotonin syndrome behaviors in a dose-dependent manner, blocked by WAY 100635 (1 mg/kg), in mice of all three genotypes, confirming the role of Htr1a receptors. The current data document markedly enhanced behavioral sensitivity to serotonin-enhancing drugs in SERT-deficient mice. These studies also show that the exaggerated behavioral responses observed in SERT +/- and -/- mice are mediated by postsynaptic Htr1a receptors, and suggest intact postsynaptic Htr1a function in SERT -/- mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0115
MDL 11939, ≥98% (HPLC)
C20H25NO