EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Asymmetric activation of tropos 2,2'-biphenol with cinchonine generates an effective catalyst for the asymmetric strecker reaction of N-tosyl-protected aldimines and ketoimines.
PMID 17768752