EMAIL THIS PAGE TO A FRIEND
Epilepsy research. Supplement
Stiripentol.
PMID 1777073
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6826
Stiripentol, ≥98% (HPLC)
C14H18O3