EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
A new family of histamine H3 receptor antagonists based on a natural product: discovery, SAR, and properties of the series.
PMID 17806176
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0095
Conessine, ≥97% (HPLC)
C24H40N2