EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
A study of the effects of p,p'-DDE and other related chlorinated hydrocarbons on inhibition of platelet aggregation.
PMID 1781738
Abstract
A series of chlorinated hydrocarbons of interest in environmental toxicology were investigated concerning their effects on human platelet aggregation. Most potent in inhibiting platelet aggregation were p,p'-DDE and Arochlor 1242. Aggregation induced by arachidonic acid (1 mM) was more sensitive to inhibition by p,p'-DDE than was aggregation induced by ADP (10 microM). The former was completely inhibited by p,p'-DDE at a concentration of 1 x 10(-4) M, whereas there was only a 31% inhibition of the latter. Addition of Ca2+ (1 mM) blocked the inhibitory effect of p,p'-DDE on aggregation induced by both arachidonic acid and ADP. Calmodulin (1 microgram/ml) blocked the inhibitory effect of p,p'-DDE on aggregation induced by arachidonic acid but not that induced by ADP. The calmodulin inhibitory drugs trifluoperazine and calmidazolium had no effect at all or only a weak effect (-14%), respectively, on platelet aggregation. Increasing the concentrations of p,p'-DDE and Arochlor 1242 caused a delay in the onset of aggregation induced by the addition of arachidonic acid. The synthesis of thromboxane B2 and other prostaglandins in platelet membranes was dose-dependently reduced by p,p'-DDE. The structurally closely related isomers o,p'-DDE and p,p'-DDT did not significantly inhibit arachidonic acid-induced platelet aggregation or thromboxane B2 synthesis. It is concluded that p,p'-DDE and Arochlor 1242 inhibited platelet aggregation by inhibiting platelet cyclooxygenase activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35484
4,4′-DDA, PESTANAL®, analytical standard
C14H10Cl2O2
100870
Bis(4-chlorophenyl)acetic acid, 98%
C14H10Cl2O2