EMAIL THIS PAGE TO A FRIEND
Journal of computational chemistry
Theoretical elucidation of the rhodium-catalyzed [4 + 2] annulation reactions.
PMID 17849391
Abstract
The reaction mechanism of the Rh-catalyzed [4 + 2] annulation of 4-alkynals with isocyanates is unraveled using density functional calculations. The reaction mechanisms of the model system and the real substituted system have been investigated and the results are compared. From our theoretical results based on the model and real substituted system, it is shown that (a) the rate-determining step is the Rh-H addition to the alkyne, (b) the formation of the cyclopentenone G and glutarimide K represents a severe competition, and (c) the product selectivity should be controlled by the amount of the isocyanates. In addition, it is demonstrated that there exist steric effects in the real substituted system, but missed in model system. Our calculations also show that although the results obtained on the model system could explain the mechanism in principle, the real substituted system could reflect the mechanism more exactly and make the reaction proceed with regioselectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178098
Glutarimide, 98%
C5H7NO2