EMAIL THIS PAGE TO A FRIEND
Current opinion in chemical biology
LSD1 and the chemistry of histone demethylation.
PMID 17851108
Abstract
The recent discovery that histone demethylation can be catalyzed by the flavin-dependent amine oxidase LSD1 has ushered in a new chapter in the chromatin-remodeling community. Herein, we discuss the rapid progress of the histone demethylase field including the recent identification of the non-heme iron-dependent histone demethylases (JmjC family), the basis for LSD1 substrate site specificity and the newly emerging potential for inhibition of these enzymes in structural and functional analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3501055
Anti-KDM1B antibody produced in rabbit, affinity isolated antibody