EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
A self-assembled nanofiber catalyst for ester hydrolysis.
PMID 17854188
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209562
2,4-Dinitrophenylacetic acid, 95%
C8H6N2O6