EMAIL THIS PAGE TO A FRIEND
Farmakologiia i toksikologiia
[Diethylnicotinamid (cordiamine) stabilization of the liver hydroxylating function in rabbits with CCl4 poisoning].
PMID 1786835
Abstract
Three and ten days after the administration of CCl4 (subcutaneously, once, 4 ml/kg of 50% oil solution) there were found a decrease of the rate of antipyrine elimination (intravenously, 50 mg/kg) from the blood plasma, an increase of the total bilirubin content, ALT activity and stimulation of lipid peroxidation processes. Cordiamine administration (subcutaneously, twice a day, 3 and 10 days) exerts the normalizing effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72372
N,N-Diethylnicotinamide, tested according to Ph.Eur.
C10H14N2O
D98807
N,N-Diethylnicotinamide, 99%
C10H14N2O