EMAIL THIS PAGE TO A FRIEND
Journal of clinical psychopharmacology
Resolution of tardive dyskinesia after addition of aripiprazole to haloperidol depot.
PMID 17873695
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0100100 Haloperidol decanoate, European Pharmacopoeia (EP) Reference Standard
C31H41ClFNO3