EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Enantioselective synthesis of (-)-cis-clavicipitic acid.
PMID 17877402
Abstract
An enantioselective synthetic method for (-)-cis-clavicipitic acid (1) was reported. 1 was obtained in 10 steps (99% ee and 20% overall yield) from 1H-indole-3-carboxylic acid methyl ester (9) via asymmetric phase-transfer catalytic alkylation and diastereoselective Pd(II)-catalyzed intramolecular aminocyclization as key steps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284734
Indole-3-carboxylic acid, ReagentPlus®, 99%
C9H7NO2