EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Racemic and optically active alpha-tocopherol analogues with a modified side chain: synthesis and biological activity].
PMID 17886430
Abstract
Information on the synthesis and biological activity of natural and synthetic analogues of alpha-tocopherol with a modified side chain is systematized. These compounds are of interest as vitamin E metabolites, hydrophilic antioxidants, and precursors of drugs with combined pharmacological properties useful in therapy of pathological disorders caused by oxidative stress.