EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
ipso-substitution: a general biochemical and biodegradation mechanism to cleave alpha-quaternary alkylphenols and bisphenol A.
PMID 17886831
Abstract
Sphingobium xenophagum Bayram is capable of metabolizing 4-alkoxyphenols and endocrine disruptive alpha-quaternary 4-nonylphenols by an ipso-substitution mechanism that involves ring hydroxylation at the site of the substituent. Here, we show that Bayram's ipso-hydroxylating activity was able to transform also bisphenol A (= dimethyl-4,4'-methylenediphenol; BPA) and 4-isopropylphenol. We identified six metabolites when resting cells of strain Bayram were incubated with BPA. They were unambiguously characterized by HPLC-UV, HPLC/MS, and HPLC/MS/MS as hydroquinone, 2-(4-hydroxyphenyl)isopropanol, 4-isopropenylphenol, 4-isopropylphenol, 4-hydroxy-4-isopropenylcyclohexa-2,5-dien-1-one, and 4-hydroxy-4-isopropylcyclohexa-2,5-dien-1-one. In experiments with 4-isopropylphenol as a substrate, 4-hydroxy-4-isopropylcyclohexa-2,5-dien-1-one, one of the metabolites from BPA, accumulated to a high degree. We could rationalize the formation of all metabolites by invoking ipso-hydroxylation and ipso-substitution mechanisms. On closer view, also classical bacterial metabolism of BPA can be well rationalized by an ipso-substitution mechanism, albeit with ipso-attack of an internal alkyl radical instead of an activated oxygen species. This highlights the important role of ipso-substitution as a versatile degradative principle utilized by diverse organisms to degrade alpha-quaternary 4-nonylphenols, 4-alkoxyphenols, and BPA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175404
4-Isopropylphenol, 98%
C9H12O