EMAIL THIS PAGE TO A FRIEND
VASA. Supplementum
[Double-blind study of the effect of various intra-arterial/intravenous prostaglandin E1 doses on hemodynamics and prostaglandin E1 metabolites].
PMID 1788751
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0286
13,14-Dihydro-15-ketoprostaglandin E2 solution, ~90%
C20H32O5