EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Sequential regimens for Helicobacter pylori eradication.
PMID 17889226
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
141615
4-Nitroimidazole, 97%
C3H3N3O2