EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
Role of 'B-b' knob-hole interactions in fibrin binding to adsorbed fibrinogen.
PMID 17892530
Abstract
The formation of a fibrin clot is supported by multiple interactions, including those between polymerization knobs 'A' and 'B' exposed by thrombin cleavage and polymerization holes 'a' and 'b' present in fibrinogen and fibrin. Although structural studies have defined the 'A-a' and 'B-b' interactions in part, it has not been possible to measure the affinities of individual knob-hole interactions in the absence of the other interactions occurring in fibrin. We designed experiments to determine the affinities of knob-hole interactions, either 'A-a' alone or 'A-a' and 'B-b' together. We used surface plasmon resonance to measure binding between adsorbed fibrinogen and soluble fibrin fragments containing 'A' knobs, desA-NDSK, or both 'A' and 'B' knobs, desAB-NDSK. The desA- and desAB-NDSK fragments bound to fibrinogen with statistically similar K(d)'s of 5.8 +/- 1.1 microm and 3.7 +/- 0.7 microm (P = 0.14), respectively. This binding was specific, as we saw no significant binding of NDSK, which has no exposed knobs. Moreover, the synthetic 'A' knob peptide GPRP and synthetic 'B' knob peptides GHRP and AHRPY, inhibited the binding of desA- and/or desAB-NDSK. The peptide inhibition findings show both 'A-a' and 'B-b' interactions participate in desAB-NDSK binding to fibrinogen, indicating 'B-b' interactions can occur simultaneously with 'A-a'. Furthermore, 'A-a' interactions are much stronger than 'B-b' because the affinity of desA-NDSK was not markedly different from desAB-NDSK.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3379
Fibrinopeptide B human, ≥97% (HPLC)
C66H93N19O25
G1895
Gly-Pro-Arg-Pro, ≥97% (HPLC)
C18H31N7O5