EMAIL THIS PAGE TO A FRIEND
Neurobiology of Learning and Memory
Treadmill exercise enhances passive avoidance learning in rats: the role of down-regulated serotonin system in the limbic system.
PMID 17892954