EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Interference by Mes [2-(4-morpholino)ethanesulfonic acid] and related buffers with phenolic oxidation by peroxidase.
PMID 17893045
Abstract
While characterizing the kinetic parameters of apoplastic phenolic oxidation by peroxidase, we found anomalies caused by the Mes [2-(4-morpholino)ethanesulfonic acid] buffer being used. In the presence of Mes, certain phenolics appeared not to be oxidized by peroxidase, yet the oxidant, H(2)O(2), was utilized. This anomaly seems to be due to the recycling of the phenolic substrate. The reaction is relatively inefficient, but at buffer concentrations of 10 mM or greater the recycling effect is nearly 100% with substrate concentrations less than 100 microM. The recycling effect is dependent on substrate structure, occurring with 4'-hydroxyacetophenone but not with 3',5'-dimethoxy-4'-hydroxyacetophenone (acetosyringone). Characterization of the reaction parameters suggests that the phenoxyl radical from the peroxidase reaction interacts with Mes, causing the reduction and regeneration of the phenol. Similar responses occurred with related buffers such as Hepes [4-(2-hydroxyethyl)piperazine-1-ethanesulfonic acid] and Pipes [piperazine-1,4-bis(2-ethanesulfonic acid)]. Results from this work and other reports in the literature indicate that great care is required in interpreting any results involving these buffers under oxidizing conditions.