EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Cardiovascular responses to noradrenaline microinjection in the ventrolateral periaqueductal gray of unanesthetized rats.
PMID 17893924
Abstract
The periaqueductal gray area (PAG) is a mesencephalic area involved in cardiovascular modulation. Noradrenaline (NA), a neurotransmitter involved in central blood pressure control, is present in the rat PAG. We report here on the cardiovascular effects caused by NA microinjection into the ventrolateral PAG (vlPAG) of unanesthetized rats and the peripheral mechanism involved in their mediation. NA microinjection in the vlPAG of unanesthetized rats evoked dose-related pressor and bradycardiac responses. No significant cardiovascular responses were observed in urethane-anesthetized rats. The pressor response was potentiated by pretreatment with the ganglion blocker pentolinium (5 or 10 mg/kg, intravenously). Pretreatment with the vasopressin antagonist dTyr(CH2)5 (Me)AVP (50 microg/kg, intravenously) blocked the pressor response evoked by the NA microinjection into the vlPAG. Additionally, circulating vasopressin content was found to be significantly increased after NA microinjection in the vlPAG. The results suggest that activation of noradrenergic synapses within the vlPAG modulates vasopressin release in unanesthetized rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V2255
[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-me-Tyr2, Arg8]-Vasopressin, ≥97% (HPLC)
C52H74N14O12S2