EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis and characterization of 5,10,15,20-tetra[3-(3-trifluoromethyl)phenoxy] porphyrin.
PMID 17909494
Abstract
The newly synthesized 5,10,15,20-tetra[3-(3-trifluoromethyl)phenoxy]porphyrin, TTFMPP, has been characterized using mass spectroscopy, 1H-, 13C- and 19F-NMR, MALDI-TOF mass spectrometry, UV-Vis and fluorescence spectrophotometry, and cyclic voltammetry. The NMR confirmed the structure of the compound and the mass spectrum was in agreement with the proposed molecular formula. The UV-Vis absorption spectrum of TTFMPP shows characteristic spectral patterns similar to those of tetraphenyl porphryin, with a Soret band at 419 nm and four Q bands at 515, 550, 590, and 648 nm. Protonation of the porphyrin with TFA resulted in the expected red shift of the Soret band. Excitation at 419 nm gave an emission at 650 nm. The quantum yield of the porphyrin was determined to be 0.08. Cyclic voltammetry was used to determine the oxidation and reduction potentials of the new porphyrin. Two quasi-reversible one-electron reductions at -1.00 and -1.32 V and a quasi-reversible oxidation at 1.20 V versus the silver/silver chloride reference electrode with tetrabutylammonium tetrafluoroborate as the supporting electrolyte in methylene chloride were observed.
Related Materials