EMAIL THIS PAGE TO A FRIEND
Biophysical chemistry
Insertion of a magnesium(II)-octacarboranyl(hexylsulfanyl) porphyrazine into liposomes: a physico-chemical study.
PMID 17916402
Abstract
The synthesis, characterization and liposome insertion of a novel magnesium(II) carboranyl-porphyrazine, i.e. [2,3,7,8,12,13,17,18-octakis-(1,2-dicarba-closo-dodecaboranyl)-hexylthio-5,10,15,20-porphyrazine]magnesium(II) complex, MgHECSPz, are described. MgHECSPz was designed to improve the potentiality in multiple approach anticancer therapy. Liposomal formulations with different surface charge were prepared as delivering agents. The obtained loaded vectors were characterized by DLS, SAXS, SANS and zeta potential measurements in order to define the overall properties and structural details of loaded liposomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
38510
Dimercaptomaleonitrile disodium salt hydrate, ≥95.0% (calc. based on dry substance, NT)
C4N2Na2S2 · xH2O