EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Increased Goodpasture antigen-binding protein expression induces type IV collagen disorganization and deposit of immunoglobulin A in glomerular basement membrane.
PMID 17916599
Abstract
Increased expression of Goodpasture antigen-binding protein (GPBP), a protein that binds and phosphorylates basement membrane collagen, has been associated with immune complex-mediated pathogenesis. However, recent reports have questioned this biological function and proposed that GPBP serves as a cytosolic ceramide transporter (CERT(L)). Thus, the role of GPBP in vivo remains unknown. New Zealand White (NZW) mice are considered healthy animals although they convey a genetic predisposition for immune complex-mediated glomerulonephritis. Here we show that NZW mice developed age-dependent lupus-prone autoimmune response and immune complex-mediated glomerulonephritis characterized by elevated GPBP, glomerular basement membrane (GBM) collagen disorganization and expansion, and deposits of IgA on disrupted GBM. Transgenic overexpression of human GPBP (hGPBP) in non-lupus-prone mice triggered similar glomerular abnormalities including deposits of IgA on a capillary GBM that underwent dissociation, in the absence of an evident autoimmune response. We provide in vivo evidence that GPBP regulates GBM collagen organization and its elevated expression causes dissociation and subsequent accumulation of IgA on the GBM. Finally, we describe a previously unrecognized pathogenic mechanism that may be relevant in human primary immune complex-mediated glomerulonephritis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB769
Anti-Collagen Type IV Antibody, Chemicon®, from goat