EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
High-activity, single-site mesoporous Pd/Al2O3 catalysts for selective aerobic oxidation of allylic alcohols.
PMID 17918275
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C86006
Crotyl alcohol, mixture of cis and trans, 96%
C4H8O