EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on flavonoids of Eupatorium odoratum L].
PMID 17918432
Abstract
To investigate the flavonoid constituents from aerial parts of Eupatorium odoratum L. systematically. The constituents of the EtoAc-soluble portions of the 95% ethanol extraction were isolated and purified by silica gel and sephadex LH-20 chromatography. Their structures were elucidated on the basis of physicochemical properties and spectral analysis. Eight compounds were isolated and identified as kaempferol-3-methoxy (I), rhamnetin (II), tamarixetin (III), quercetin (IV), kaempferol (V), apigenin (VI), luteolin (VII), dihydrokaempferide (VIII). Compound I , II and VI were isolated for the first time from genus Eupatorium, and VIII which is rare flavonoid was obtainted for the first time from family Compositae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17799
Rhamnetin, analytical standard
C16H12O7