EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Hypoalgesia in mice lacking GABA transporter subtype 1.
PMID 17918738
Abstract
gamma-Aminobutyric acid (GABA) transporters play a key role in the regulation of GABA neurotransmission. We reported previously that overexpression of the GABA transporter subtype 1 (GAT1), the major form of the GABA transporter in the CNS, led to hyperalgesia in mice. In the present study, nociceptive responses of GAT1-knockout mice (GAT1(-/-)) were compared with those of heterozygous (GAT(+/-)) and wild-type (GAT(+/+)) mice by four conventional pain models (tail-immersion test, hot-plate test, acetic acid-induced abdominal constriction test, and formalin test). In addition, the analgesic effects of two GAT1-selective inhibitors, NO-711 and tiagabine, were examined in all three genotypes using the same four models. Our data demonstrated that GAT1 deficiency because of genetic knockout or acute blockade by selective inhibitors leads to hypoalgesia in mice. These results confirmed the crucial role of GAT1 in the regulation of nociceptive threshold and suggested that GAT1 inhibitors have the potential for clinical use in pain therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N142
NO-711 hydrochloride, ≥98% (HPLC)
C21H22N2O3 · HCl