EMAIL THIS PAGE TO A FRIEND
Cell biology international
Effect of nicotine and polyaromtic hydrocarbons on cerebral endothelial cells.
PMID 17920942
Abstract
The present study was designed to investigate the effect of nicotine and polyaromatic hydrocarbon compounds on cerebral endothelial cells (CECs). Nicotine treatments from 15 min to 5h did not cause any changes in the expression and localization of principal junctional proteins. One day of treatment with a relatively high concentration of nicotine induced a decrease in the expression of the tight junction protein ZO-1, occludin, and the adherens junction protein, cadherin. Treatment with 3 x 10(-5)M phenanthrene for 24h caused a redistribution of occludin from the Triton X-100 insoluble to the Triton X-100 soluble fraction. Transendothelial electrical resistance was not significantly affected by 24h treatments with nicotine, methylanthracene or phenanthrene. However, 24h nicotine treatment increased transendothelial permeability in CECs exposed to oxidative stress. Both nicotine and phenanthrene were able to regulate the expression of a large number of proteins as revealed by 2D electrophoresis. Our experiments suggest that tobacco smoking may affect the junctional complex of CECs, and that this effect is enhanced by oxidative stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5439 2,3-Dimethoxy-1,4-naphthoquinone, ≥99%, solid
C12H10O4