EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
B:b interactions are essential for polymerization of variant fibrinogens with impaired holes 'a'.
PMID 17922804
Abstract
Fibrin polymerization is mediated by interactions between knobs 'A' and 'B' exposed by thrombin cleavage, and holes 'a' and 'b' always present in fibrinogen. The role of A:a interactions is well established, but the roles of knob:hole interactions A:b, B:b or B:a remain ambiguous. To determine whether A:b or B:b interactions have a role in thrombin-catalyzed polymerization, we examined a series of fibrinogen variants with substitutions altering holes 'a': gamma364Ala, gamma364His or gamma364Val. We examined thrombin- and reptilase-catalyzed fibrinopeptide release by high-performance liquid chromatography, fibrin clot formation by turbidity, fibrin clot structure by scanning electron microscopy (SEM) and factor (F) XIIIa-catalyzed crosslinking by sodium dodecylsulfate polyacrylamide gel electrophoresis. Thrombin-catalyzed fibrinopeptide A release was normal, but fibrinopeptide B release was delayed for all variants. The variant fibrinogens all showed markedly impaired thrombin-catalyzed polymerization; polymerization of gamma364Val and gamma364His were more delayed than gamma364Ala. There was absolutely no polymerization of any variant with reptilase, which exposed only knobs 'A'. SEM showed that the variant clots formed after 24 h had uniform, ordered fibers that were thicker than normal. Polymerization of the variant fibrinogens was inhibited dose-dependently by the addition of either Gly-Pro-Arg-Pro (GPRP) or Gly-His-Arg-Pro (GHRP), peptides that specifically block holes 'a' and 'b', respectively. FXIIIa-catalyzed crosslinking between gamma-chains was markedly delayed for all the variants. These results demonstrate that B:b interactions are critical for polymerization of variant fibrinogens with impaired holes 'a'. Based on these data, we propose a model wherein B:b interactions participate in protofibril formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3379
Fibrinopeptide B human, ≥97% (HPLC)
C66H93N19O25
G1895
Gly-Pro-Arg-Pro, ≥97% (HPLC)
C18H31N7O5