EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and anti-mycobacterial activity of (E)-N'-(monosubstituted-benzylidene)isonicotinohydrazide derivatives.
PMID 17923172
Abstract
A series of 22 (E)-N'-(monosubstituted-benzylidene)isonicotinohydrazide derivatives have been synthesized and evaluated for their in vitro antibacterial activity against Mycobacterium tuberculosis H(37)Rv using Alamar Blue susceptibility test and the activity expressed as the minimum inhibitory concentration (MIC) in mug/mL. Compounds 2f, 2g, 2j, 2k and 2q exhibited a significant activity (0.31-0.62 microg/mL) when compared with first line drugs such as isoniazid (INH) and rifampicin (RIP) and could be a good starting point to develop new lead compounds in the fight against multi-drug resistant tuberculosis.