EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design of a potent reactivator of tabun-inhibited acetylcholinesterase--synthesis and evaluation of (E)-1-(4-carbamoylpyridinium)-4-(4-hydroxyiminomethylpyridinium)-but-2-ene dibromide (K203).
PMID 17924614
Abstract
Acetylcholinesterase reactivators are crucial antidotes for the treatment of organophosphate intoxication. Among the organophosphates, with the exception of soman, tabun (GA) intoxications are the least responsive to treatment with commercially available therapeutics. A rational design was used to increase reactivation ability and decrease the toxicity of the novel reactivator. (E)-1-(4-carbamoylpyridinium)-4-(4-hydroxyiminomethylpyridinium)-but-2-ene dibromide (K203) has better properties than previously tested compounds in vitro and, therefore, is a potential candidate for the treatment of GA intoxication in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9053
Pyridine-2-aldoxime methochloride
C7H9N2O · Cl