EMAIL THIS PAGE TO A FRIEND
Redox report : communications in free radical research
Quantitative determination of ortho- and meta-tyrosine as biomarkers of protein oxidative damage in beta-thalassemia.
PMID 17925094
Abstract
Oxidative stress in thalassemia is caused by secondary iron overload and stems from blood transfusion and increased iron uptake. In this study, we hypothesized that levels of o- and m-tyrosine, products of hydroxyl radical attack on phenylalanine, would be elevated in beta-thalassemia (intermediate). This study represents the first report in which specific markers of protein oxidative damage have been quantified in thalassemia. We used GC/MS to assay o- and m-tyrosine at the femtomole level using only a few microliters of plasma. Levels of both markers were significantly higher in patients with beta-thalassemia than in controls and were positively correlated with serum ferritin, malondialdehyde, superoxide dismutase, glutathione peroxidase and glutathione. We conclude that o- and m-tyrosine are useful biomarkers of oxidative damage to proteins in thalassemia (intermediate) and may also be useful markers in other iron overload diseases. Positive correlations between o- and m-tyrosine levels and malondialdehyde as well as antioxidants such as superoxide dismutase, glutathione peroxidase and glutathione, are indicative of the broad impact of oxidative stress on blood plasma in thalassemia, with up-regulation of antioxidant proteins probably reflecting a homeostatic response to these increased stress levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3629
DL-m-Tyrosine, crystalline
C9H11NO3
93851
DL-o-Tyrosine, ≥96.0% (NT)
C9H11NO3